Order Entry
Finland
ContactUsLinkComponent
6-Chloro-7-methylchromone ≥98.0% (by GC)
Irritant
6-Chloro-7-methylchromone ≥98.0% (by GC)
Catalog # TCIAC3035-1G
Supplier:  TCI
CAS Number:  
Irritant
6-Chloro-7-methylchromone ≥98.0% (by GC)
Catalog # TCIAC3035-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Purity (GC):
    min. 98.0 %
  • Melting point:
    169.0 to 173.0 deg-C

Specification Test Results

About this item

 :
 :194.62 g/mol
 :GHS07
CAS Number:67029-84-9
Formula:C₁₀H₇ClO₂
MDL Number:MFCD00239401