Order Entry
Finland
ContactUsLinkComponent
 
 
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl ≥98.0% (by GC)
 :  TCI
 
 
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +42.0 to +46.0 deg (neat)