Order Entry
Finland
ContactUsLinkComponent
 
5-CFDA (5-Carboxyfluorescein diacetate) ≥93.0% (by HPLC, titration analysis)
 :  TCI
 
5-CFDA (5-Carboxyfluorescein diacetate) ≥93.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 93.0 area%
  • Purity (Neutralization titration):
    min. 93.0 %