7-(Diethylamino)-3-(1-methyl-2-benzimidazolyl)coumarin ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:41044-12-6
:
Formula:C₂₁H₂₁N₃O₂
MDL Number:MFCD00051349
:347.42 g/mol
EINECS:255-186-8
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:221.0 to 225.0 deg-C
- Lambda max.:413.0 to 415.0 nm (EtOH)