2-((4-Cyanophenyl)amino)acetic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
About this item
Synonyms: N-(4-CN-Phenyl)-Gly-OH, N-(4-Cyanophenyl)glycine
- CAS Number:42288-26-6
- Formula:C₉H₈N₂O₂
- MDL Number:MFCD01464022
- Molecular Weight:176.1700 g/mol
- EINECS:459-560-3
2 Options Available Below
Product Details & Documents
Synonyms: N-(4-CN-Phenyl)-Gly-OH, N-(4-Cyanophenyl)glycine
- CAS Number:42288-26-6
- Formula:C₉H₈N₂O₂
- MDL Number:MFCD01464022
- Molecular Weight:176.1700 g/mol
- EINECS:459-560-3
Specifications for Product Family
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.TCIAC2771-5G - Specifications:
More
Specifications
Cat. No.TCIAC2771-25G
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



