
2-Bromo-3'-chloro-4'-fluoroacetophenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:63529-30-6
:
Formula:C₈H₅BrClFO
MDL Number:MFCD05154991
:251.48 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:34.0 to 38.0 deg-C