5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:27006-76-4
:
Formula:C₉H₁₁ClN₂O
MDL Number:MFCD00052538
:158.59 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:77.0 to 80.0 deg-C