Order Entry
Finland
ContactUsLinkComponent
1-Cyclohexenylboronic acid pinacol ester ≥95.0% (by GC)
1-Cyclohexenylboronic acid pinacol ester ≥95.0% (by GC)
Catalog # TCIAC2707-1G
Supplier:  TCI
CAS Number:  
1-Cyclohexenylboronic acid pinacol ester ≥95.0% (by GC)
Catalog # TCIAC2707-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :208.1049 g/mol
Flash Pt:103°C
Storage Temperature:Refrigerator
CAS Number:141091-37-4
Formula:C₁₂H₂₁BO₂
MDL Number:MFCD05663845