N-Carbobenzoxy-D-2-phenylglycine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:17609-52-8
:MFCD00064977
Formula:C₁₆H₁₅NO₄
MDL Number:MFCD00021703
:285.29 g/mol
EINECS:241-582-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 95.0 %
- Melting point:130.0 to 135.0 deg-C
- Specific rotation [a]20/D:-114.0 to -118.0 deg (C=1, ethanol)