Order Entry
Finland
ContactUsLinkComponent
 
 
 
2-(Chloromethyl)-1,2-epoxybutane ≥95.0% (by GC)
  TCIAC2429-5G
 :  TCI
 :  
 
 
2-(Chloromethyl)-1,2-epoxybutane ≥95.0% (by GC)
  TCIAC2429-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

 

 

 :
 :120.58 g/mol
 :137°C
 :Refrigerator
 :2920
 :8,II
 :GHS02
 :GHS05
 :75484-32-1
 :C₅H₉ClO