Chitosan, 20-100 mPa.s (0.5% in 0.5% Acetic acid, 20 °C)
: TCI
:9012-76-4
:MFCD00149018
:MFCD00161512
:222-311-2
- Appearance:White to Almost white powder to crystaline
- Deacetylation value:min. 80.0 % (calcd.on dried substance)
- Drying loss:max. 12.0 %
- Viscosity:20 to 100mPa-s (0.5% in 0.5%Acetic acid soln., 20deg-C) (after drying)