Order Entry
Finland
ContactUsLinkComponent
1,3,5-Cyclohexanetricarbonitrile (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)
Toxic
1,3,5-Cyclohexanetricarbonitrile (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)
Supplier:  TCI
Certificates
Toxic
1,3,5-Cyclohexanetricarbonitrile (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 % (total of isomer)

Specification Test Results

Frequently Bought Together