cis-1,2-Cyclohexanedimethanol ≥97.0% (by GC)
: TCI
:15753-50-1
:
:C₈H₁₆O₂
:MFCD00062987
:144.21 g/mol
:112°C
:239-843-6
- Appearance:White or Colorless to Yellow powder to lump to clear liquid
- Purity (GC):min. 97.0 %
- Melting point:38.0 to 43.0 deg-C