N-Carbobenzoxy-L-threonine benzyl ester ≥98.0% (by HPLC, total nitrogen)
TCIAC2285-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:80.0 to 84.0 deg-C
- Specific rotation [a]20/D:-9.0 to -13.0 deg (C=8, CHCl3)
:
:343.38 g/mol
:16597-50-5
:C₁₉H₂₁NO₅
:MFCD00077041