Coumarin 314 ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAC2268-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:148.0 to 152.0 deg-C
Specification Test Results
About this item
:
:313.35 g/mol
EINECS:259-825-1
CAS Number:55804-66-5
Formula:C₁₈H₁₉NO₄
MDL Number:MFCD00051334