
Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:94790-35-9
:207915-99-9
Formula:C₅H₁₂ClF₆N₂P
MDL Number:MFCD01862891
:280.58 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %