Order Entry
Finland
ContactUsLinkComponent
 
Chloromethyl polystyrene resin 1.8 - 2.5 mmol/g, cross-linked with 2% DVB (100-200 mesh)
Catalog # TCIAC1747-5G
Supplier:  TCI
CAS Number:  
Chloromethyl polystyrene resin 1.8 - 2.5 mmol/g, cross-linked with 2% DVB (100-200 mesh)
Catalog # TCIAC1747-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Loading:
    1.8 to 2.5 mmol/g

Specification Test Results

About this item

 :
CAS Number:55844-94-5
MDL Number:MFCD00146406