Order Entry
Finland
ContactUsLinkComponent
 
 
N-Carbobenzoxy-1,5-diaminopentane hydrochloride ≥98.0% (by titrimetric analysis)
 :  TCI
 
 
N-Carbobenzoxy-1,5-diaminopentane hydrochloride ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    170.0 to 174.0 deg-C