

Cyclopropyldiphenylsulphonium tetrafluoroborate ≥95.0% (by titrimetric analysis)
Catalog # TCIAC1390-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Ion exchange titration):min. 95.0 %
- Melting point:139.0 to 143.0 deg-C
Specification Test Results
About this item
:
:314.15 g/mol
EINECS:251-530-6
UN:1759
ADR:8,III
:GHS05
:GHS07
CAS Number:33462-81-6
Formula:C₁₅H₁₅BF₄S
MDL Number:MFCD00011873