Order Entry
Finland
ContactUsLinkComponent
 
Kalsiumasetaatti-xH2O 99.0-100.5% (by anhydrous basis), Puriss., meets analytical specification of FCC, E263, Fluka™
 
Kalsiumasetaatti-xH2O 99.0-100.5% (by anhydrous basis), Puriss., meets analytical specification of FCC, E263, Fluka™
 

 

  • Assay (calculated on the anhydrous basis):
    99.0 - 100.5 %
  • Water (Karl Fischer):
    Max. 7.0 %
  • Water insoluble matter:
    Max. 0.3 %
  • pH (10 %, 20°C):
    7 - 9.0
  • Arsenic (As):
    Max. 1 ppm
  • Copper (Cu):
    Max. 0.001 %
  • Iron (Fe):
    Max. 0.002 %
  • Mercury (Hg):
    Max. 1 ppm
  • Lead (Pb):
    Max. 2 ppm
  • Zinc (Zn):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.03 %
  • Fluoride (F):
    Max. 0.005 %
  • Sulfate (SO₄):
    Max. 0.05 %
  • Formic acid, formiates other impurities (HCOOH):
    Max. 1000 ppm

 

 

Frequently Bought Together