
Cinnamyl bromide ≥98.0% (by GC, titration analysis)
Catalog # TCIAC1109-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
About this item
:
:197.08 g/mol
Boiling Pt:107°C
Storage Temperature:Refrigerator
EINECS:224-511-5
UN:3261
ADR:8,III
:GHS05
CAS Number:4392-24-9
Formula:C₉H₉Br
MDL Number:MFCD00000245