Chloroform-[D₁] stabilised
Supplier: TCI
About this item
Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D
- CAS Number:865-49-6
- Formula:CDCl₃
- MDL Number:MFCD00000827
- Molecular Weight:120.38 g/mol
- Boiling Pt:61°C
- EINECS:212-742-4
- UN:1888
- ADR:6.1,III
- :Stabilised with silver
2 Options Available Below
Product Details & Documents
Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D
- CAS Number:865-49-6
- Formula:CDCl₃
- MDL Number:MFCD00000827
- Molecular Weight:120.38 g/mol
- Boiling Pt:61°C
- EINECS:212-742-4
- UN:1888
- ADR:6.1,III
- :Stabilised with silver
Documents
Manual Bioscience and Analyticalscience
Käyttöohje
Specifications for Product Family
Specification Test Results
- Appearance:Colorless clear liquid
- Degree of deuteration:min. 99.6 atom%D
Product Family Options
Product Information
Suositukset
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



