Diethyl-1,2-cyclohexanedicarboxylate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:10138-59-7
:
Formula:C₁₂H₂₀O₄
MDL Number:MFCD00029449
:228.29 g/mol
Boiling Pt:135°C
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %