
Sodium-3-methylsalicylate ≥97.0% (by HPLC, titration analysis)
TCIAC0411-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Ion exchange titration):min. 97.0 % (calcd.on dried substance)
- Solubility in Water:almost transparency
- Drying loss:max. 5.0 %
:
:174.13 g/mol
:251-202-2
:GHS07
:32768-20-0
:C₈H₈O₃
:MFCD00058985