2-[2-[(tert-Butoxycarbonyl)amino]thiazol-4-yl]acetic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:89336-46-9
:
Formula:C₁₀H₁₄N₂O₄S
MDL Number:MFCD02682394
:258.29 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:166.0 to 170.0 deg-C
- Solubility in Methanol:almost transparency