1,4-Benzenediboronic acid bis(neopentyl glycol) ester ≥98.0% (by GC, titration analysis)
Catalog # TCIAB6050-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g/mol
- Seq:9220
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:233.0 to 237.0 deg-C
Specification Test Results
About this item
:
:301.99 g/mol
CAS Number:5565-36-6
Formula:C₁₆H₂₄B₂O₄
MDL Number:MFCD01075726