

1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAB6045-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
Specification Test Results
About this item
:
:394.26 g/mol
UN:1759
ADR:8,II
:GHS05
:GHS07
CAS Number:245679-18-9
Formula:C₂₁H₂₇BF₄N₂
MDL Number:MFCD02684542