
1-Boc-3,5-dimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
: TCI
:1073354-70-7
:
:C₁₆H₂₇BN₂O₄
:MFCD09027070
:322.2076 g/mol
:GHS07
:Refrigerator
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:109.0 to 113.0 deg-C