Order Entry
Finland
ContactUsLinkComponent
2-[[tert-Butyl(dimethyl)silyl]oxy]ethylamine ≥95.0% (by GC, titration analysis)
Irritant
2-[[tert-Butyl(dimethyl)silyl]oxy]ethylamine ≥95.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
2-[[tert-Butyl(dimethyl)silyl]oxy]ethylamine ≥95.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %
  • Purity (Nonaqueous Titration):
    min. 95.0 %

Specification Test Results