
3-Bromobenzenesulphonamide ≥98.0% (by GC)
: TCI
:89599-01-9
:
:C₆H₆BrNO₂S
:MFCD00084903
:236.09 g/mol
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:156.0 to 160.0 deg-C
Frequently Bought Together