Order Entry
Finland
ContactUsLinkComponent
1-Benzyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
Irritant
1-Benzyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Irritant
1-Benzyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results