Order Entry
Finland
ContactUsLinkComponent
Benzyl(ethyl)dimethylammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
Irritant
Benzyl(ethyl)dimethylammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB5427-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Benzyl(ethyl)dimethylammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB5427-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Ion exchange titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :444.41 g/mol
 :GHS07
CAS Number:1186103-43-4
Formula:C₁₃H₁₈F₆N₂O₄S₂
MDL Number:MFCD18251282