
2-Bromo-4-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:328-89-2
:
Formula:C₈H₄BrF₃O₂
MDL Number:MFCD07368837
:269.02 g/mol
EINECS:206-339-2
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %