
Bis(4-bromophenyl)iodonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:139139-81-4
:
Formula:C₁₃H₈Br₂F₃IO₃S
MDL Number:MFCD21608478
:587.97 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 %
- Melting point:210.0 to 214.0 deg-C