
4-Butoxy-2,3-difluorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:156487-12-6
:
Formula:C₁₀H₁₃BF₂O₃
MDL Number:MFCD19441186
:230.02 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 109.0 %