
3-Bromo-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:352535-97-8
:
Formula:C₆H₅BBrFO₂
MDL Number:MFCD05664228
:218.816 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):97.0 to 109.0 %
Specification Test Results
Frequently Bought Together