Order Entry
Finland
ContactUsLinkComponent
trans,trans-4-n-Butyl-4'-(3,4-difluorophenyl)bicyclohexyl ≥98.0% (by GC)
trans,trans-4-n-Butyl-4'-(3,4-difluorophenyl)bicyclohexyl ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
trans,trans-4-n-Butyl-4'-(3,4-difluorophenyl)bicyclohexyl ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    44.0 to 49.0 deg-C
  • Mesomorphic range:
    44.0 to 123.0 deg-C

Specification Test Results

Frequently Bought Together