
(2,2'-Bipyridine)bis(2-phenylpyridinato)iridium(III) Hexafluorophosphate ≥90.0% (by HPLC)
Catalog # TCIAB4893-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 90.0 area%
- Elemental analysis (Nitrogen):6.00 to 8.00 %
Specification Test Results
About this item
:
:801.75 g/mol
Melting Pt:250°C
:GHS07
CAS Number:106294-60-4
Formula:C₃₂H₂₄F₆IrN₄P
MDL Number:MFCD28976160