
10-Bromo-7H-benzo[c]carbazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1698-16-4
:
Formula:C₁₆H₁₀BrN
MDL Number:MFCD22054961
:296.17 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light gray to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:127.0 to 131.0 deg-C