
3-Bromo-4-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:101420-81-9
:
Formula:C₇H₄BrNO₄
MDL Number:MFCD04117949
:246.02 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:201.0 to 205.0 deg-C