
5-Bromo-1,2,3-trimethoxybenzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2675-79-8
:
Formula:C₉H₁₁BrO₃
MDL Number:MFCD00017169
:247.09 g/mol
Boiling Pt:273°C
EINECS:220-223-9
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:78.0 to 82.0 deg-C