
3-Bromobenzylamine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10269-01-9
:
Formula:C₇H₈BrN
MDL Number:MFCD01026119
Boiling Pt:245°C
UN:2735
ADR:8,III
:186.05 g/mol
:GHS05
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together