
4-Bromothiazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:34259-99-9
:
Formula:C₃H₂BrNS
MDL Number:MFCD06657592
:164.03 g/mol
Boiling Pt:190°C
Flash Pt:101°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %