Anthracene-9,10-diboronic acid bis(pinacol) ester ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:863992-56-7
:
Formula:C₂₆H₃₂B₂O₄
MDL Number:MFCD16294538
:430.16 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %