1,3-Bis(4-chlorophenyl)thiourea ≥98.0% (by HPLC)
Catalog # TCIAB4481-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 96.0 %
- Melting point:169.0 to 173.0 deg-C
Specification Test Results
About this item
:
:297.20 g/mol
CAS Number:1220-00-4
Formula:C₁₃H₁₀Cl₂N₂S
MDL Number:MFCD00275761