
1-Boc-4-(aminomethyl)piperidine ≥98.0% (by GC, titration analysis)
: TCI
:144222-22-0
:
:C₁₁H₂₂N₂O₂
:MFCD01076207
:214.31 g/mol
:110°C
:GHS07
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together