N-(tert-Butoxycarbonyl)-4-phenyl-L-phenylalanine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB4289-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+20.0 ~ +23.0 deg (C=2, MeOH)
Specification Test Results
About this item
:
:341.41 g/mol
CAS Number:147923-08-8
Formula:C₂₀H₂₃NO₄
MDL Number:MFCD00191184