Order Entry
Finland
ContactUsLinkComponent
BIX 01294 trihydrochloride, hydrate ≥85.0% (by total nitrogen basis)
BIX 01294 trihydrochloride, hydrate ≥85.0% (by total nitrogen basis)
Catalog # TCIAB4211-25MG
Supplier:  TCI
CAS Number:  
BIX 01294 trihydrochloride, hydrate ≥85.0% (by total nitrogen basis)
Catalog # TCIAB4211-25MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 85.0 % (calcd.on anh.substance)
  • Water:
    2.0 to 7.0 %

Specification Test Results

About this item

 :
 :600.03 g/mol (anhy)
Storage Temperature:Refrigerator
CAS Number:1392399-03-9
Formula:C₂₈H₃₈N₆O₂·3HCl
MDL Number:MFCD16618384