
(R)-1,2-Butanediol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:40348-66-1
:
Formula:C₄H₁₀O₂
MDL Number:MFCD09953765
:90.12 g/mol
Boiling Pt:60°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Specific rotation [a]20/D:+15.0 to +18.0 deg (C=3、EtOH)