ω-Bromo-3-cyanoacetophenone ≥98.0% (by GC)
TCIAB4079-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:69.0 to 73.0 deg-C
:
:224.06 g/mol
:2923
:8,III
:GHS05
:GHS06
:50916-55-7
:C₉H₆BrNO
:MFCD00833267